Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134.2 | NP_001125.1 | 1 - 2 | 128 | 143 |
GenBank mRNA | AK297669.1 | - | 1 - 2 | 126 | 143 |
AK297674.1 | - | 1 - 2 | 123 | 143 | |
AK314817.1 | - | 1 - 2 | 122 | 143 | |
BC027881.1 | - | 1 - 2 | 114 | 143 | |
V01514.1 | - | 1 - 2 | 128 | 143 |