Assay Design



Gene Transcripts

Gene Symbol
AFP
Entrez Gene ID
174
Gene Name
alpha fetoprotein
Gene Aliases
AFPD, FETA, HPAFP
Chromosome Location
Chr.4: 73436219 - 73455785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134.2 NP_001125.1 4 - 5 521 93
GenBank mRNA AK314817.1 - 4 - 5 515 93
BC027881.1 - 4 - 5 507 93
V01514.1 - 4 - 5 521 93

View Full Product Details