Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014244.4 | NP_055059.2 | 20 - 21 | 3192 | 104 |
XM_017010063.1 | XP_016865552.1 | 19 - 20 | 2628 | 104 | |
GenBank mRNA | AJ003125.1 | - | 20 - 21 | 3090 | 104 |
BX479109.1 | - | 2 - 3 | 155 | 104 |