Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085049.2 | NP_001078518.1 | 1 - 2 | 103 | 134 |
XM_005247229.2 | XP_005247286.1 | 1 - 2 | 292 | 134 | |
GenBank mRNA | AF043938.1 | - | 1 - 2 | 86 | 134 |
BC047690.1 | - | 1 - 2 | 96 | 134 | |
BI546150.1 | - | 1 - 2 | 300 | 134 | |
CN356600.1 | - | 1 - 2 | 103 | 134 |