Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006334.3 | NP_006325.1 | 1 - 2 | 543 | 69 |
NM_014279.4 | NP_055094.1 | 1 - 2 | 557 | 69 | |
GenBank mRNA | AK289719.1 | - | 1 - 2 | 270 | 69 |
AK289921.1 | - | 1 - 2 | 152 | 69 | |
BC008763.2 | - | 1 - 2 | 279 | 69 | |
BC011741.2 | - | 1 - 2 | 279 | 69 | |
BT007146.1 | - | 1 - 2 | 63 | 69 | |
BX419781.2 | - | 1 - 2 | 474 | 69 | |
BX461034.2 | - | 1 - 2 | 525 | 69 | |
D82343.1 | - | 1 - 2 | 378 | 69 |