Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077206.3 | NP_001070674.1 | 8 - 9 | 1076 | 84 |
NM_001077207.3 | NP_001070675.1 | 8 - 9 | 1076 | 84 | |
NM_001077208.3 | NP_001070676.1 | 8 - 9 | 1076 | 84 | |
NM_001191049.2 | NP_001177978.1 | 7 - 8 | 993 | 84 | |
NM_001300744.2 | NP_001287673.1 | 8 - 9 | 1076 | 84 | |
NM_001300745.2 | NP_001287674.1 | 8 - 9 | 1076 | 84 | |
NM_001318119.1 | NP_001305048.1 | 8 - 9 | 1073 | 84 | |
NM_001318120.1 | NP_001305049.1 | 10 - 11 | 1290 | 84 | |
NM_016211.4 | NP_057295.2 | 8 - 9 | 1076 | 84 | |
GenBank mRNA | AB018358.1 | - | 8 - 9 | 932 | 84 |
AB018359.1 | - | 8 - 9 | 932 | 84 | |
AB020712.1 | - | 10 - 11 | 1257 | 84 | |
AF139184.1 | - | 7 - 8 | 879 | 84 | |
AK002161.1 | - | 8 - 9 | 949 | 84 | |
AK225934.1 | - | 8 - 9 | 936 | 84 | |
AK295810.1 | - | 7 - 8 | 891 | 84 | |
BC047883.2 | - | 8 - 9 | 980 | 84 | |
BC084583.1 | - | 8 - 9 | 915 | 84 | |
BC117221.1 | - | 8 - 9 | 944 | 84 | |
BC143489.1 | - | 8 - 9 | 941 | 84 | |
BC143491.1 | - | 8 - 9 | 944 | 84 | |
BC143492.2 | - | 8 - 9 | 944 | 84 | |
CA397330.1 | - | 4 - 5 | 423 | 84 | |
CR933696.1 | - | 8 - 9 | 949 | 84 |