Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006364.3 | NP_006355.2 | 1 - 2 | 499 | 169 |
XM_005267262.1 | XP_005267319.1 | 1 - 2 | 214 | 169 | |
XM_011536355.2 | XP_011534657.1 | 1 - 2 | 214 | 169 | |
XM_017020928.1 | XP_016876417.1 | 1 - 2 | 214 | 169 | |
GenBank mRNA | AK299862.1 | - | 1 - 2 | 183 | 169 |
AK303485.1 | - | 1 - 2 | 213 | 169 | |
BC036649.1 | - | 1 - 2 | 212 | 169 | |
BM541968.1 | - | 1 - 2 | 197 | 169 | |
DC335582.1 | - | 1 - 2 | 499 | 169 | |
X97064.1 | - | 1 - 2 | 140 | 169 |