Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006364.3 | NP_006355.2 | 5 - 6 | 1126 | 128 |
XM_005267262.1 | XP_005267319.1 | 5 - 6 | 841 | 128 | |
XM_011536355.2 | XP_011534657.1 | 5 - 6 | 841 | 128 | |
XM_017020928.1 | XP_016876417.1 | 5 - 6 | 841 | 128 | |
GenBank mRNA | AK293944.1 | - | 4 - 5 | 431 | 128 |
AK299862.1 | - | 4 - 5 | 723 | 128 | |
AK303485.1 | - | 3 - 4 | 534 | 128 | |
AK312259.1 | - | 5 - 6 | 708 | 128 | |
BC036649.1 | - | 5 - 6 | 839 | 128 | |
X97064.1 | - | 5 - 6 | 767 | 128 |