Assay Design



Gene Transcripts

Gene Symbol
CRISP1
Entrez Gene ID
167
Gene Name
cysteine rich secretory protein 1
Gene Aliases
AEGL1, ARP, CRISP-1, HEL-S-57, HSCRISP1D, HSCRISP1G, HUMARP
Chromosome Location
Chr.6: 49834257 - 49877096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001131.2 NP_001122.2 6 - 7 606 109
NM_001205220.1 NP_001192149.1 6 - 7 617 109
XM_011514315.2 XP_011512617.1 3 - 4 737 109
XM_017010320.1 XP_016865809.1 3 - 4 555 109
XM_017010321.1 XP_016865810.1 3 - 4 265 109
GenBank mRNA AK292497.1 - 6 - 7 587 109
D38451.1 - 7 - 8 606 109
GQ472203.1 - 6 - 7 600 109
S80310.1 - 7 - 8 606 109
X95237.1 - 6 - 7 600 109

View Full Product Details