Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304743.1 | NP_001291672.1 | 5 - 6 | 1151 | 95 |
NM_014720.3 | NP_055535.2 | 5 - 6 | 1151 | 95 | |
XM_011540401.2 | XP_011538703.1 | 5 - 6 | 1163 | 95 | |
GenBank mRNA | AB002804.1 | - | 5 - 6 | 822 | 95 |
AF273048.1 | - | 5 - 6 | 624 | 95 | |
AK302855.1 | - | 5 - 6 | 584 | 95 | |
AK307389.1 | - | 6 - 7 | 703 | 95 | |
BC047762.1 | - | 5 - 6 | 1144 | 95 | |
BC047885.1 | - | 5 - 6 | 1151 | 95 | |
BC064804.1 | - | 5 - 6 | 1147 | 95 | |
BC111565.1 | - | 5 - 6 | 621 | 95 | |
D86959.1 | - | 5 - 6 | 1101 | 95 |