Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144578.3 | NP_653179.1 | 1 - 2 | 165 | 60 |
XM_017021789.1 | XP_016877278.1 | 1 - 2 | 201 | 60 | |
GenBank mRNA | AK025580.1 | - | 1 - 2 | 128 | 60 |
AK315501.1 | - | 1 - 2 | 162 | 60 | |
AL834285.1 | - | 1 - 2 | 148 | 60 | |
BC015621.2 | - | 1 - 2 | 126 | 60 |