Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145138.1 | NP_001138610.1 | 7 - 8 | 797 | 71 |
NM_001243984.1 | NP_001230913.1 | 7 - 8 | 806 | 71 | |
NM_001243985.1 | NP_001230914.1 | 7 - 8 | 806 | 71 | |
NM_021975.3 | NP_068810.3 | 7 - 8 | 806 | 71 | |
GenBank mRNA | AK303262.1 | - | 7 - 8 | 785 | 71 |
AK308020.1 | - | 7 - 8 | 750 | 71 | |
AK308478.1 | - | 6 - 7 | 852 | 71 | |
BC033522.1 | - | 6 - 7 | 663 | 71 | |
BC110830.1 | - | 7 - 8 | 729 | 71 | |
L19067.1 | - | 6 - 7 | 665 | 71 | |
M62399.1 | - | 7 - 8 | 745 | 71 |