Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015302.1 | NP_056117.1 | 6 - 7 | 535 | 75 |
XM_011526684.2 | XP_011524986.1 | 5 - 6 | 422 | 75 | |
GenBank mRNA | AB020648.1 | - | 6 - 7 | 508 | 75 |
BC013947.2 | - | 6 - 7 | 507 | 75 | |
BC064390.1 | - | 6 - 7 | 535 | 75 | |
BC157882.1 | - | 6 - 7 | 526 | 75 |