Assay Design



Gene Transcripts

Gene Symbol
HAUS5
Entrez Gene ID
23354
Gene Name
HAUS augmin like complex subunit 5
Gene Aliases
KIAA0841, dgt5
Chromosome Location
Chr.19: 35612744 - 35625349 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015302.1 NP_056117.1 9 - 10 751 102
XM_011526684.2 XP_011524986.1 8 - 9 638 102
GenBank mRNA AB020648.1 - 9 - 10 724 102
BC013947.2 - 9 - 10 723 102
BC064390.1 - 9 - 10 751 102
BC157882.1 - 9 - 10 742 102
BC171905.1 - 9 - 10 743 102

View Full Product Details