Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015155.2 | NP_055970.1 | 4 - 5 | 610 | 93 |
XM_005252432.3 | XP_005252489.1 | 4 - 5 | 619 | 93 | |
XM_005252435.3 | XP_005252492.1 | 4 - 5 | 403 | 93 | |
XM_011519435.2 | XP_011517737.1 | 4 - 5 | 454 | 93 | |
XM_017015987.1 | XP_016871476.1 | 3 - 4 | 2659 | 93 | |
XM_017015988.1 | XP_016871477.1 | 3 - 4 | 2657 | 93 | |
XM_017015989.1 | XP_016871478.1 | 3 - 4 | 2659 | 93 | |
XM_017015990.1 | XP_016871479.1 | 4 - 5 | 2178 | 93 | |
XM_017015991.1 | XP_016871480.1 | 3 - 4 | 2673 | 93 | |
XM_017015993.1 | XP_016871482.1 | 3 - 4 | 338 | 93 | |
XM_017015994.1 | XP_016871483.1 | 4 - 5 | 375 | 93 | |
GenBank mRNA | BC131630.1 | - | 3 - 4 | 327 | 93 |
BC152443.1 | - | 3 - 4 | 334 | 93 | |
D86971.2 | - | 3 - 4 | 334 | 93 |