Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004958.3 | NP_004949.1 | 17 - 18 | 2772 | 59 |
XM_005263438.2 | XP_005263495.1 | 17 - 18 | 2779 | 59 | |
XM_011541166.2 | XP_011539468.1 | 17 - 18 | 2772 | 59 | |
XM_017000900.1 | XP_016856389.1 | 14 - 15 | 2235 | 59 | |
XM_017000901.1 | XP_016856390.1 | 15 - 16 | 2523 | 59 | |
XM_017000902.1 | XP_016856391.1 | 17 - 18 | 2772 | 59 | |
GenBank mRNA | AB209995.1 | - | 17 - 18 | 2753 | 59 |
AK302863.1 | - | 17 - 18 | 2764 | 59 | |
AK304273.1 | - | 17 - 18 | 2761 | 59 | |
BC117166.1 | - | 16 - 17 | 2669 | 59 | |
L34075.1 | - | 17 - 18 | 2730 | 59 | |
U88966.1 | - | 17 - 18 | 2730 | 59 |