Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004958.3 | NP_004949.1 | 27 - 28 | 4227 | 89 |
XM_005263438.2 | XP_005263495.1 | 27 - 28 | 4234 | 89 | |
XM_011541166.2 | XP_011539468.1 | 27 - 28 | 4227 | 89 | |
XM_017000900.1 | XP_016856389.1 | 24 - 25 | 3690 | 89 | |
XM_017000901.1 | XP_016856390.1 | 25 - 26 | 3978 | 89 | |
XM_017000902.1 | XP_016856391.1 | 27 - 28 | 4227 | 89 | |
GenBank mRNA | AB209995.1 | - | 27 - 28 | 4208 | 89 |
AK304273.1 | - | 27 - 28 | 4216 | 89 | |
BC117166.1 | - | 26 - 27 | 4124 | 89 | |
L34075.1 | - | 27 - 28 | 4185 | 89 | |
U88966.1 | - | 27 - 28 | 4182 | 89 |