Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004958.3 | NP_004949.1 | 49 - 50 | 7051 | 77 |
XM_005263438.2 | XP_005263495.1 | 49 - 50 | 7058 | 77 | |
XM_017000900.1 | XP_016856389.1 | 46 - 47 | 6514 | 77 | |
XM_017000901.1 | XP_016856390.1 | 47 - 48 | 6802 | 77 | |
GenBank mRNA | AB209995.1 | - | 49 - 50 | 7032 | 77 |
AK126762.1 | - | 11 - 12 | 2347 | 77 | |
BC117166.1 | - | 48 - 49 | 6948 | 77 | |
L34075.1 | - | 49 - 50 | 7009 | 77 | |
U88966.1 | - | 49 - 50 | 7006 | 77 |