Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130146.1 | NP_001123618.1 | 10 - 11 | 1293 | 91 |
NM_001130147.1 | NP_001123619.1 | 10 - 11 | 1320 | 91 | |
NM_001130148.1 | NP_001123620.1 | 9 - 10 | 1266 | 91 | |
NM_032358.3 | NP_115734.1 | 11 - 12 | 1347 | 91 | |
GenBank mRNA | AK027638.1 | - | 11 - 12 | 1265 | 91 |
AK293165.1 | - | 10 - 11 | 1231 | 91 | |
AK308216.1 | - | 10 - 11 | 1275 | 91 | |
AK316029.1 | - | 9 - 10 | 1268 | 91 | |
BC006444.2 | - | 10 - 11 | 1236 | 91 |