Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005692.4 | NP_005683.2 | 9 - 10 | 1386 | 78 |
NM_007189.2 | NP_009120.1 | 9 - 10 | 1386 | 78 | |
XM_017011639.1 | XP_016867128.1 | 9 - 10 | 1263 | 78 | |
XM_017011640.1 | XP_016867129.1 | 9 - 10 | 1348 | 78 | |
GenBank mRNA | AF261091.1 | - | 9 - 10 | 1243 | 78 |
AF261092.1 | - | 9 - 10 | 1224 | 78 | |
AJ005016.1 | - | 7 - 8 | 913 | 78 | |
AL050291.1 | - | 8 - 9 | 1178 | 78 | |
BC001661.2 | - | 9 - 10 | 1218 | 78 | |
BG720644.1 | - | 4 - 5 | 524 | 78 | |
BT007451.1 | - | 8 - 9 | 1132 | 78 |