Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145664.1 | NP_001139136.1 | 5 - 6 | 469 | 147 |
XM_011511771.2 | XP_011510073.1 | 6 - 7 | 1007 | 147 | |
XM_011511772.1 | XP_011510074.1 | 8 - 9 | 681 | 147 | |
XM_011511773.1 | XP_011510075.1 | 4 - 5 | 352 | 147 | |
XM_011511776.1 | XP_011510078.1 | 7 - 8 | 640 | 147 | |
XM_011511777.1 | XP_011510079.1 | 2 - 3 | 1010 | 147 | |
XM_011511778.1 | XP_011510080.1 | 2 - 3 | 347 | 147 | |
XM_017004851.1 | XP_016860340.1 | 8 - 9 | 691 | 147 | |
XM_017004852.1 | XP_016860341.1 | 5 - 6 | 478 | 147 | |
XM_017004853.1 | XP_016860342.1 | 8 - 9 | 691 | 147 | |
XM_017004854.1 | XP_016860343.1 | 8 - 9 | 691 | 147 | |
GenBank mRNA | AK124976.1 | - | 4 - 5 | 412 | 147 |
AK131289.1 | - | 8 - 9 | 706 | 147 | |
AK298610.1 | - | 5 - 6 | 469 | 147 |