Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052886.2 | NP_443118.1 | 3 - 4 | 559 | 86 |
XM_011516807.2 | XP_011515109.1 | 2 - 3 | 343 | 86 | |
GenBank mRNA | AK172820.1 | - | 3 - 4 | 475 | 86 |
AK312030.1 | - | 3 - 4 | 466 | 86 | |
AY007723.1 | - | 3 - 4 | 536 | 86 | |
BC012367.1 | - | 3 - 4 | 559 | 86 | |
CV574204.1 | - | 3 - 4 | 559 | 86 |