Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143821.2 | NP_001137293.2 | 18 - 19 | 2525 | 64 |
NM_001256470.1 | NP_001243399.1 | 21 - 22 | 2660 | 64 | |
NM_001256787.1 | NP_001243716.1 | 15 - 16 | 2216 | 64 | |
NM_019012.5 | NP_061885.2 | 16 - 17 | 2351 | 64 | |
XM_005253400.1 | XP_005253457.1 | 16 - 17 | 2311 | 64 | |
XM_006719091.1 | XP_006719154.1 | 17 - 18 | 2365 | 64 | |
XM_006719093.1 | XP_006719156.1 | 15 - 16 | 2227 | 64 | |
XM_006719096.1 | XP_006719159.1 | 20 - 21 | 2602 | 64 | |
XM_011520714.1 | XP_011519016.1 | 19 - 20 | 2566 | 64 | |
XM_011520715.1 | XP_011519017.1 | 18 - 19 | 2485 | 64 | |
XM_011520716.1 | XP_011519018.1 | 20 - 21 | 2602 | 64 | |
XM_011520717.1 | XP_011519019.1 | 18 - 19 | 2485 | 64 | |
XM_017019495.1 | XP_016874984.1 | 20 - 21 | 2602 | 64 | |
XM_017019496.1 | XP_016874985.1 | 20 - 21 | 2602 | 64 | |
XM_017019497.1 | XP_016874986.1 | 18 - 19 | 2416 | 64 | |
XM_017019498.1 | XP_016874987.1 | 15 - 16 | 2183 | 64 | |
XM_017019499.1 | XP_016874988.1 | 16 - 17 | 2311 | 64 | |
GenBank mRNA | AB051473.2 | - | 18 - 19 | 2421 | 64 |
AB642244.1 | - | 21 - 22 | 2626 | 64 | |
AF302150.1 | - | 16 - 17 | 2249 | 64 | |
AK001529.1 | - | 2 - 3 | 255 | 64 | |
AK023127.1 | - | 4 - 5 | 372 | 64 | |
AK056054.1 | - | 9 - 10 | 1177 | 64 | |
AK294739.1 | - | 16 - 17 | 2217 | 64 | |
AK295151.1 | - | 21 - 22 | 2584 | 64 | |
AK296277.1 | - | 14 - 15 | 2128 | 64 | |
AK297391.1 | - | 14 - 15 | 1992 | 64 | |
AL834259.1 | - | 2 - 3 | 209 | 64 | |
BC004164.2 | - | 7 - 8 | 1036 | 64 | |
BC013133.2 | - | 5 - 6 | 483 | 64 | |
BC127091.1 | - | 15 - 16 | 2176 | 64 | |
BC127092.1 | - | 16 - 17 | 2249 | 64 |