Assay Design



Gene Transcripts

Gene Symbol
TPT1
Entrez Gene ID
7178
Gene Name
tumor protein, translationally-controlled 1
Gene Aliases
HRF, TCTP, p02, p23
Chromosome Location
Chr.13: 45333471 - 45341284 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286272.1 NP_001273201.1 1 - 2 250 95
NM_003295.3 NP_003286.1 1 - 2 250 95
GenBank mRNA AF072098.1 - - 130 95
AK130847.1 - 1 - 2 149 95
AK296587.1 - 1 - 2 149 95
AK296952.1 - 1 - 2 149 95
AK312951.1 - 1 - 2 149 95
AY117678.1 - 1 - 2 150 95
BC003352.2 - 1 - 2 133 95
BC012431.1 - 1 - 2 109 95
BC052333.1 - 1 - 2 139 95
BP215975.1 - 1 - 2 250 95
X16064.1 - 1 - 2 129 95
X64899.1 - 1 - 2 122 95

View Full Product Details