Assay Design



Gene Transcripts

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
Chromosome Location
Chr.11: 6621324 - 6655849 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003737.3 NP_003728.1 15 - 16 6771 71
GenBank mRNA AB053446.1 - 15 - 16 6774 71
AL831828.1 - 2 - 3 593 71

View Full Product Details