Assay Design



Gene Transcripts

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
Chromosome Location
Chr.11: 6621324 - 6655849 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003737.3 NP_003728.1 18 - 19 7272 119
GenBank mRNA AB053446.1 - 18 - 19 7275 119
AL831828.1 - 5 - 6 1094 119

View Full Product Details