Assay Design



Gene Transcripts

Gene Symbol
SLC15A5
Entrez Gene ID
729025
Gene Name
solute carrier family 15 member 5
Gene Aliases
-
Chromosome Location
Chr.12: 16188485 - 16277685 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170798.1 NP_001164269.1 1 - 2 362 111

View Full Product Details