Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040274.2 | NP_001035364.2 | 23 - 24 | 2247 | 164 |
GenBank mRNA | AK128130.1 | - | 23 - 24 | 2301 | 164 |
AM293674.1 | - | 23 - 24 | 1966 | 164 | |
BC012225.1 | - | 13 - 14 | 1144 | 164 |