Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303270.1 | NP_001290199.1 | 12 - 13 | 1840 | 75 |
NM_020877.3 | NP_065928.2 | 10 - 11 | 2968 | 75 | |
XM_011523663.1 | XP_011521965.1 | 12 - 13 | 1836 | 75 | |
XM_011523664.2 | XP_011521966.1 | 13 - 14 | 1985 | 75 | |
XM_011523666.1 | XP_011521968.1 | 12 - 13 | 1836 | 75 | |
XM_011523667.2 | XP_011521969.1 | 11 - 12 | 1739 | 75 | |
XM_011523668.1 | XP_011521970.1 | 2 - 3 | 219 | 75 | |
XM_011523670.2 | XP_011521972.1 | 12 - 13 | 1836 | 75 | |
XM_017024219.1 | XP_016879708.1 | 12 - 13 | 1836 | 75 | |
GenBank mRNA | AB040936.2 | - | 10 - 11 | 2968 | 75 |
AK292607.1 | - | 12 - 13 | 1836 | 75 | |
BC034225.1 | - | 12 - 13 | 1839 | 75 |