Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020877.3 | NP_065928.2 | 21 - 22 | 4967 | 89 |
XM_011523663.1 | XP_011521965.1 | 23 - 24 | 3835 | 89 | |
XM_011523664.2 | XP_011521966.1 | 24 - 25 | 3984 | 89 | |
XM_011523666.1 | XP_011521968.1 | 23 - 24 | 3835 | 89 | |
XM_011523667.2 | XP_011521969.1 | 22 - 23 | 3738 | 89 | |
XM_011523668.1 | XP_011521970.1 | 13 - 14 | 2218 | 89 | |
XM_011523669.2 | XP_011521971.1 | 11 - 12 | 1902 | 89 | |
XM_011523670.2 | XP_011521972.1 | 23 - 24 | 3835 | 89 | |
XM_017024218.1 | XP_016879707.1 | 10 - 11 | 1919 | 89 | |
XM_017024219.1 | XP_016879708.1 | 23 - 24 | 3835 | 89 | |
GenBank mRNA | AB040936.2 | - | 21 - 22 | 4967 | 89 |