Assay Design



Gene Transcripts

Gene Symbol
DNAH2
Entrez Gene ID
146754
Gene Name
dynein axonemal heavy chain 2
Gene Aliases
DNAHC2, DNHD3
Chromosome Location
Chr.17: 7717354 - 7833744 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020877.3 NP_065928.2 43 - 44 8204 68
XM_011523663.1 XP_011521965.1 45 - 46 7072 68
XM_011523664.2 XP_011521966.1 46 - 47 7221 68
XM_011523666.1 XP_011521968.1 45 - 46 7072 68
XM_011523667.2 XP_011521969.1 44 - 45 6975 68
XM_011523668.1 XP_011521970.1 35 - 36 5455 68
XM_011523669.2 XP_011521971.1 33 - 34 5139 68
XM_011523670.2 XP_011521972.1 45 - 46 7072 68
XM_017024218.1 XP_016879707.1 32 - 33 5156 68
XM_017024219.1 XP_016879708.1 45 - 46 7072 68
GenBank mRNA AB040936.2 - 43 - 44 8204 68

View Full Product Details