Assay Design



Gene Transcripts

Gene Symbol
DNAH2
Entrez Gene ID
146754
Gene Name
dynein axonemal heavy chain 2
Gene Aliases
DNAHC2, DNHD3
Chromosome Location
Chr.17: 7717354 - 7833744 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020877.3 NP_065928.2 59 - 60 10641 59
XM_011523663.1 XP_011521965.1 61 - 62 9509 59
XM_011523664.2 XP_011521966.1 62 - 63 9658 59
XM_011523666.1 XP_011521968.1 60 - 61 9320 59
XM_011523667.2 XP_011521969.1 60 - 61 9412 59
XM_011523668.1 XP_011521970.1 51 - 52 7892 59
XM_011523669.2 XP_011521971.1 49 - 50 7576 59
XM_011523670.2 XP_011521972.1 61 - 62 9509 59
XM_017024218.1 XP_016879707.1 48 - 49 7593 59
XM_017024219.1 XP_016879708.1 61 - 62 9509 59
GenBank mRNA AB040936.2 - 59 - 60 10641 59

View Full Product Details