Assay Design



Gene Transcripts

Gene Symbol
EIF2A
Entrez Gene ID
83939
Gene Name
eukaryotic translation initiation factor 2A
Gene Aliases
CDA02, EIF-2A, MST089, MSTP004, MSTP089
Chromosome Location
Chr.3: 150546678 - 150586016 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319043.1 NP_001305972.1 5 - 6 528 139
NM_001319045.1 NP_001305974.1 6 - 7 588 139
NM_032025.4 NP_114414.2 6 - 7 603 139
XM_011513224.1 XP_011511526.1 6 - 7 586 139
GenBank mRNA AF212241.1 - 6 - 7 552 139
AF497978.1 - 6 - 7 481 139
AK027356.1 - 6 - 7 494 139
AK298586.1 - 5 - 6 439 139
AL833101.1 - 6 - 7 487 139
BC011885.1 - 6 - 7 484 139
BX647951.1 - 6 - 7 473 139

View Full Product Details