Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319043.1 | NP_001305972.1 | 7 - 8 | 746 | 116 |
NM_001319044.1 | NP_001305973.1 | 6 - 7 | 638 | 116 | |
NM_001319045.1 | NP_001305974.1 | 8 - 9 | 806 | 116 | |
NM_032025.4 | NP_114414.2 | 8 - 9 | 821 | 116 | |
XM_011513224.1 | XP_011511526.1 | 8 - 9 | 804 | 116 | |
GenBank mRNA | AF172818.1 | - | 7 - 8 | 689 | 116 |
AF212241.1 | - | 8 - 9 | 770 | 116 | |
AF497978.1 | - | 8 - 9 | 699 | 116 | |
AK027356.1 | - | 8 - 9 | 712 | 116 | |
AK293993.1 | - | 6 - 7 | 529 | 116 | |
AK298586.1 | - | 7 - 8 | 657 | 116 | |
AL833101.1 | - | 8 - 9 | 705 | 116 | |
BC011885.1 | - | 8 - 9 | 702 | 116 | |
BX647951.1 | - | 8 - 9 | 691 | 116 |