Assay Design
Gene Transcripts
- Gene Symbol
- C8orf44
- Entrez Gene ID
-
56260
- Gene Name
- chromosome 8 open reading frame 44
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 66667552 - 66681149 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_019607.2 |
NP_062553.1 |
1 - 2 |
155 |
129 |
GenBank mRNA |
BC014448.1 |
- |
1 - 2 |
89 |
129 |
View Full Product Details