Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002180.2 | NP_002171.2 | 6 - 7 | 1012 | 138 |
XM_005273974.2 | XP_005274031.1 | 6 - 7 | 976 | 138 | |
XM_005273976.1 | XP_005274033.1 | 6 - 7 | 1012 | 138 | |
XM_017017669.1 | XP_016873158.1 | 5 - 6 | 868 | 138 | |
XM_017017670.1 | XP_016873159.1 | 2 - 3 | 773 | 138 | |
XM_017017671.1 | XP_016873160.1 | 6 - 7 | 1012 | 138 | |
GenBank mRNA | AB208802.1 | - | - | 954 | 138 |
AK312565.1 | - | 6 - 7 | 970 | 138 | |
BC000290.2 | - | 6 - 7 | 925 | 138 | |
BC025299.1 | - | 6 - 7 | 957 | 138 | |
BC080519.1 | - | 6 - 7 | 957 | 138 | |
BC105088.1 | - | 6 - 7 | 949 | 138 | |
BC105090.1 | - | 6 - 7 | 949 | 138 |