Assay Design
Gene Transcripts
- Gene Symbol
- GPR32
- Entrez Gene ID
-
2854
- Gene Name
- G protein-coupled receptor 32
- Gene Aliases
-
RVDR1
- Chromosome Location
-
Chr.19: 50770464 - 50771732 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001506.2 |
NP_001497.1 |
1 - 1 |
1191 |
72 |
GenBank mRNA |
BC095544.1 |
- |
1 - 1 |
1192 |
72 |
View Full Product Details