Assay Design



Gene Transcripts

Gene Symbol
TSPAN19
Entrez Gene ID
144448
Gene Name
tetraspanin 19
Gene Aliases
-
Chromosome Location
Chr.12: 85014311 - 85036277 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100917.1 NP_001094387.1 7 - 8 133
XM_005268669.4 XP_005268726.1 7 - 8 133
XM_017018862.1 XP_016874351.1 6 - 7 133
XM_017018863.1 XP_016874352.1 6 - 7 133
GenBank mRNA CB852874.1 - 2 - 3 336 133
CB854430.1 - 2 - 3 336 133

View Full Product Details