Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020778.4 | NP_065829.3 | 9 - 10 | 4897 | 94 |
XM_017022444.1 | XP_016877933.1 | 8 - 9 | 4895 | 94 | |
GenBank mRNA | AB037751.1 | - | 4 - 5 | 1846 | 94 |
AL832372.1 | - | 4 - 5 | 2134 | 94 | |
AL833276.1 | - | 4 - 5 | 2213 | 94 | |
AL833288.1 | - | 5 - 6 | 3579 | 94 | |
AY044449.1 | - | 9 - 10 | 4897 | 94 |