Assay Design



Gene Transcripts

Gene Symbol
SLC2A8
Entrez Gene ID
29988
Gene Name
solute carrier family 2 member 8
Gene Aliases
GLUT8, GLUTX1
Chromosome Location
Chr.9: 127397138 - 127407898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271711.1 NP_001258640.1 6 - 7 956 98
NM_001271712.1 NP_001258641.1 4 - 5 586 98
NM_014580.4 NP_055395.2 6 - 7 956 98
NR_073416.1 3 - 4 389 98
XM_006717084.3 XP_006717147.1 6 - 7 951 98
XM_011518602.2 XP_011516904.1 6 - 7 872 98
XM_011518603.2 XP_011516905.1 4 - 5 611 98
XM_011518604.2 XP_011516906.1 4 - 5 535 98
XM_017014653.1 XP_016870142.1 4 - 5 576 98
GenBank mRNA AF289587.1 - 6 - 7 864 98
AJ245937.1 - 6 - 7 866 98
BC019043.2 - 6 - 7 908 98
BM044230.1 - 3 - 4 290 98
Y17801.1 - 6 - 7 889 98

View Full Product Details