Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163735.1 | NP_001157207.1 | 14 - 15 | 2224 | 103 |
NM_025109.5 | NP_079385.2 | 14 - 15 | 2224 | 103 | |
XM_011525286.1 | XP_011523588.1 | 17 - 18 | 1911 | 103 | |
XM_011525287.1 | XP_011523589.1 | 17 - 18 | 1905 | 103 | |
XM_011525290.2 | XP_011523592.1 | 15 - 16 | 1773 | 103 | |
XM_017025157.1 | XP_016880646.1 | 17 - 18 | 1885 | 103 | |
XM_017025158.1 | XP_016880647.1 | 16 - 17 | 1818 | 103 | |
XM_017025159.1 | XP_016880648.1 | 15 - 16 | 1779 | 103 | |
XM_017025160.1 | XP_016880649.1 | 17 - 18 | 1911 | 103 | |
XM_017025161.1 | XP_016880650.1 | 15 - 16 | 1703 | 103 | |
GenBank mRNA | AB209035.1 | - | 14 - 15 | 2182 | 103 |
AK293903.1 | - | 14 - 15 | 2831 | 103 | |
AK304073.1 | - | 14 - 15 | 1575 | 103 | |
BC008900.2 | - | 14 - 15 | 1615 | 103 |