Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163735.1 | NP_001157207.1 | 18 - 19 | 2789 | 69 |
XM_011525286.1 | XP_011523588.1 | 21 - 22 | 2476 | 69 | |
XM_011525287.1 | XP_011523589.1 | 21 - 22 | 2470 | 69 | |
XM_011525290.2 | XP_011523592.1 | 19 - 20 | 2338 | 69 | |
XM_017025157.1 | XP_016880646.1 | 21 - 22 | 2450 | 69 | |
XM_017025158.1 | XP_016880647.1 | 20 - 21 | 2383 | 69 | |
XM_017025159.1 | XP_016880648.1 | 19 - 20 | 2344 | 69 | |
XM_017025160.1 | XP_016880649.1 | 21 - 22 | 2476 | 69 | |
XM_017025161.1 | XP_016880650.1 | 19 - 20 | 2268 | 69 | |
GenBank mRNA | AB209035.1 | - | 18 - 19 | 2747 | 69 |
AK304073.1 | - | 18 - 19 | 2140 | 69 | |
AL133017.1 | - | 4 - 5 | 2564 | 69 |