Assay Design



Gene Transcripts

Gene Symbol
OLFM1
Entrez Gene ID
10439
Gene Name
olfactomedin 1
Gene Aliases
AMY, NOE1, NOELIN1, OlfA
Chromosome Location
Chr.9: 135075243 - 135121184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282611.1 NP_001269540.1 1 - 2 473 111
GenBank mRNA AK290478.1 - 3 - 4 261 111

View Full Product Details