Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199193.2 | NP_954663.1 | 1 - 2 | 226 | 153 |
NM_199194.2 | NP_954664.1 | 1 - 2 | 226 | 153 | |
GenBank mRNA | AF420603.1 | - | 1 - 2 | 48 | 153 |
BG422104.1 | - | 1 - 2 | 226 | 154 | |
BG422293.1 | - | 1 - 2 | 156 | ||
L38616.1 | - | 1 - 2 | 54 | 153 |