Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261840.1 | NP_001248769.1 | 3 - 4 | 458 | 81 |
NM_004899.4 | NP_004890.2 | 3 - 4 | 458 | 81 | |
NM_199191.2 | NP_954661.1 | 3 - 4 | 458 | 81 | |
NM_199192.2 | NP_954662.1 | 3 - 4 | 458 | 81 | |
NM_199193.2 | NP_954663.1 | 4 - 5 | 524 | 81 | |
NM_199194.2 | NP_954664.1 | 4 - 5 | 524 | 81 | |
GenBank mRNA | AF015767.1 | - | 3 - 4 | 333 | 81 |
AF420602.1 | - | 3 - 4 | 280 | 81 | |
AF420603.1 | - | 4 - 5 | 346 | 81 | |
AF420604.1 | - | 3 - 4 | 289 | 81 | |
AF420605.1 | - | 3 - 4 | 280 | 81 | |
AK000097.1 | - | 3 - 4 | 344 | 81 | |
AK291086.1 | - | 3 - 4 | 334 | 81 | |
AY438031.1 | - | 2 - 3 | 206 | 81 | |
BC001251.1 | - | 2 - 3 | 228 | 81 | |
BI559404.1 | - | - | 393 | 81 | |
BU172408.1 | - | - | 405 | 81 | |
L38616.1 | - | 4 - 5 | 352 | 81 |