Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267041.2 | NP_001253970.1 | 9 - 10 | 1052 | 103 |
NM_001282342.1 | NP_001269271.1 | 8 - 9 | 975 | 103 | |
NM_014154.4 | NP_054873.2 | 11 - 12 | 1221 | 103 | |
NM_015396.5 | NP_056211.2 | 11 - 12 | 1221 | 103 | |
NM_213654.3 | NP_998819.1 | 11 - 12 | 1275 | 103 | |
XM_006713564.3 | XP_006713627.1 | 10 - 11 | 1140 | 103 | |
XM_006713565.2 | XP_006713628.1 | 11 - 12 | 1181 | 103 | |
XM_006713566.3 | XP_006713629.1 | 9 - 10 | 1035 | 103 | |
XM_006713567.3 | XP_006713630.1 | 10 - 11 | 1140 | 103 | |
XM_011512632.1 | XP_011510934.1 | 10 - 11 | 1088 | 103 | |
XM_011512633.1 | XP_011510935.1 | 10 - 11 | 1142 | 103 | |
XM_017006096.1 | XP_016861585.1 | 9 - 10 | 1035 | 103 | |
XM_017006100.1 | XP_016861589.1 | 10 - 11 | 1140 | 103 | |
XM_017006101.1 | XP_016861590.1 | 10 - 11 | 1050 | 103 | |
GenBank mRNA | AF109184.1 | - | 11 - 12 | 974 | 103 |
AK001721.1 | - | 9 - 10 | 783 | 103 | |
AK001882.1 | - | 2 - 3 | 356 | 103 | |
AK098500.1 | - | 1 - 2 | 1510 | 103 | |
AK289577.1 | - | 11 - 12 | 917 | 103 | |
AK296837.1 | - | 9 - 10 | 752 | 103 | |
AK299769.1 | - | 8 - 9 | 695 | 103 | |
AK304558.1 | - | 9 - 10 | 728 | 103 | |
AK315932.1 | - | 5 - 6 | 418 | 103 | |
BC013424.1 | - | 11 - 12 | 975 | 103 | |
BC032661.1 | - | 11 - 12 | 1085 | 103 | |
BC041699.1 | - | 11 - 12 | 900 | 103 |