Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136033.2 | NP_001129505.1 | 3 - 4 | 309 | 134 |
NM_001271096.1 | NP_001258025.1 | 3 - 4 | 287 | 134 | |
NM_001271098.1 | NP_001258027.1 | 3 - 4 | 287 | 134 | |
NM_001271100.1 | NP_001258029.1 | 3 - 4 | 309 | 134 | |
NM_014281.4 | NP_055096.2 | 3 - 4 | 290 | 134 | |
NM_078480.2 | NP_510965.1 | 3 - 4 | 290 | 134 | |
XM_011516929.2 | XP_011515231.1 | 4 - 5 | 489 | 134 | |
XM_011516930.2 | XP_011515232.1 | 4 - 5 | 391 | 134 | |
XM_017013234.1 | XP_016868723.1 | 3 - 4 | 1822 | 134 | |
XM_017013235.1 | XP_016868724.1 | 3 - 4 | 392 | 134 | |
XM_017013236.1 | XP_016868725.1 | 3 - 4 | 1822 | 134 | |
XM_017013237.1 | XP_016868726.1 | 4 - 5 | 551 | 134 | |
XM_017013238.1 | XP_016868727.1 | 4 - 5 | 351 | 134 | |
XM_017013239.1 | XP_016868728.1 | 3 - 4 | 317 | 134 | |
XM_017013240.1 | XP_016868729.1 | 3 - 4 | 317 | 134 | |
GenBank mRNA | AF114818.1 | - | 3 - 4 | 271 | 134 |
AF190744.1 | - | 3 - 4 | 199 | 134 | |
AF217197.2 | - | 3 - 4 | 217 | 134 | |
AK055941.1 | - | 2 - 3 | 134 | ||
AK292373.1 | - | 3 - 4 | 271 | 134 | |
AL522753.3 | - | 3 - 4 | 237 | 134 | |
BC008875.2 | - | 3 - 4 | 256 | 134 | |
BC009734.1 | - | 3 - 4 | 239 | 134 | |
BC011265.1 | - | 3 - 4 | 226 | 134 | |
BC035435.2 | - | 2 - 3 | 190 | 134 | |
BG771821.1 | - | 3 - 4 | 308 | 134 | |
BX418925.2 | - | 3 - 4 | 239 | 134 | |
CR457143.1 | - | 3 - 4 | 207 | 134 | |
U51586.1 | - | 2 - 3 | 204 | 134 |