Assay Design



Gene Transcripts

Gene Symbol
PDCD10
Entrez Gene ID
11235
Gene Name
programmed cell death 10
Gene Aliases
CCM3, TFAR15
Chromosome Location
Chr.3: 167683891 - 167735690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011512368.2 XP_011510670.1 2 - 3 1023 120
XM_011512369.2 XP_011510671.1 2 - 3 1028 120

View Full Product Details