Assay Design



Gene Transcripts

Gene Symbol
INPP4A
Entrez Gene ID
3631
Gene Name
inositol polyphosphate-4-phosphatase type I A
Gene Aliases
INPP4, TVAS1
Chromosome Location
Chr.2: 98444592 - 98594068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134224.1 NP_001127696.1 22 - 23 2828 84
NM_001134225.1 NP_001127697.1 21 - 22 2813 84
NM_001566.2 NP_001557.1 22 - 23 2711 84
NM_004027.2 NP_004018.1 22 - 23 2711 84
XM_006712499.3 XP_006712562.1 22 - 23 2826 84
XM_006712500.3 XP_006712563.1 22 - 23 2823 84
XM_006712501.3 XP_006712564.1 21 - 22 2808 84
XM_006712502.3 XP_006712565.1 21 - 22 2793 84
XM_006712503.3 XP_006712566.1 22 - 23 2826 84
XM_006712504.3 XP_006712567.1 22 - 23 2709 84
XM_006712505.3 XP_006712568.1 22 - 23 2826 84
XM_006712506.3 XP_006712569.1 21 - 22 2676 84
XM_006712507.3 XP_006712570.1 22 - 23 2709 84
XM_006712508.3 XP_006712571.1 22 - 23 2826 84
XM_011511122.2 XP_011509424.1 21 - 22 2776 84
XM_011511123.2 XP_011509425.1 22 - 23 2826 84
XM_011511124.2 XP_011509426.1 21 - 22 2691 84
XM_011511125.2 XP_011509427.1 19 - 20 2708 84
XM_011511127.2 XP_011509429.1 10 - 11 1511 84
XM_017003993.1 XP_016859482.1 22 - 23 2550 84
XM_017003994.1 XP_016859483.1 21 - 22 2691 84
XM_017003995.1 XP_016859484.1 21 - 22 2673 84
XM_017003996.1 XP_016859485.1 22 - 23 2826 84
XM_017003997.1 XP_016859486.1 22 - 23 2768 84
XM_017003998.1 XP_016859487.1 22 - 23 2765 84
XM_017003999.1 XP_016859488.1 21 - 22 2749 84
XM_017004000.1 XP_016859489.1 21 - 22 2676 84
XM_017004001.1 XP_016859490.1 20 - 21 2658 84
XM_017004002.1 XP_016859491.1 20 - 21 2576 84
XM_017004003.1 XP_016859492.1 22 - 23 2709 84
GenBank mRNA AF368319.1 - 20 - 21 2435 84
AK307823.1 - 7 - 8 815 83
BC028361.2 - 21 - 22 2813 84
U26398.1 - 22 - 23 2612 84
U96919.1 - 22 - 23 2612 84

View Full Product Details