Assay Design



Gene Transcripts

Gene Symbol
INPP4A
Entrez Gene ID
3631
Gene Name
inositol polyphosphate-4-phosphatase type I A
Gene Aliases
INPP4, TVAS1
Chromosome Location
Chr.2: 98444592 - 98594068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134224.1 NP_001127696.1 7 - 8 865 87
NM_001134225.1 NP_001127697.1 7 - 8 865 87
NM_001566.2 NP_001557.1 7 - 8 865 87
NM_004027.2 NP_004018.1 7 - 8 865 87
XM_006712499.3 XP_006712562.1 7 - 8 860 87
XM_006712500.3 XP_006712563.1 7 - 8 860 87
XM_006712501.3 XP_006712564.1 7 - 8 860 87
XM_006712502.3 XP_006712565.1 7 - 8 860 87
XM_006712503.3 XP_006712566.1 7 - 8 860 87
XM_006712504.3 XP_006712567.1 7 - 8 860 87
XM_006712505.3 XP_006712568.1 7 - 8 860 87
XM_006712506.3 XP_006712569.1 7 - 8 860 87
XM_006712507.3 XP_006712570.1 7 - 8 860 87
XM_006712508.3 XP_006712571.1 7 - 8 860 87
XM_011511122.2 XP_011509424.1 6 - 7 810 87
XM_011511123.2 XP_011509425.1 7 - 8 860 87
XM_011511124.2 XP_011509426.1 7 - 8 860 87
XM_011511125.2 XP_011509427.1 4 - 5 742 87
XM_017003993.1 XP_016859482.1 7 - 8 701 87
XM_017003994.1 XP_016859483.1 7 - 8 860 87
XM_017003995.1 XP_016859484.1 7 - 8 860 87
XM_017003996.1 XP_016859485.1 7 - 8 860 87
XM_017003997.1 XP_016859486.1 7 - 8 919 87
XM_017003998.1 XP_016859487.1 7 - 8 919 87
XM_017003999.1 XP_016859488.1 7 - 8 918 87
XM_017004000.1 XP_016859489.1 7 - 8 860 87
XM_017004001.1 XP_016859490.1 7 - 8 860 87
XM_017004002.1 XP_016859491.1 7 - 8 778 87
XM_017004003.1 XP_016859492.1 7 - 8 860 87
GenBank mRNA AF368319.1 - 5 - 6 472 87
BC028361.2 - 7 - 8 865 87
U26398.1 - 7 - 8 766 87
U96919.1 - 7 - 8 766 87

View Full Product Details